Learn More
MyBioSource Melan-A Recombinant Protein, 0.2mg
SDP
Supplier:  MyBioSource MBS76105680.2 MG
Encompass_Preferred
MyBioSource offers recombinant Melan-A protein for research use. This is a 17.5 kDa protein expressed in E. coli. It is supplied at 0.2 mg per vial, lyophilized powder. The protein can be used for ELISA, Western Blot, and other antibody-based assays. The recommended storage temperature is -20C. This research-grade reagent allows scientists to study the biological and immunological properties of Melan-A.
Catalog No. NC2204120
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.