Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanin, MP Biomedicals™

Catalog No. ICN15534391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15534391 1 g
ICN15534380 100 mg
ICN15534383 250 mg
ICN15534394 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15534391 Supplier MP Biomedicals, Inc Supplier No. 0215534391
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8049-97-6
Molecular Formula C18H10N2O4
Molecular Weight (g/mol) 318.288
InChI Key XUMBMVFBXHLACL-UHFFFAOYSA-N
Synonym melanin, phaeomelanins, d0wq4g, melanin from sepia officinalis, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1; 2,5 .0; 13,17 .0; 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1^ 2,5 .0^ 13,17 .0^ 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone
PubChem CID 6325610
SMILES CC1=C2C3=C(C4=CNC5=C(C(=O)C(=O)C(=C45)C3=CN2)C)C(=O)C1=O

Specifications

Color Brown to Black
Quantity 1 g
Formula Weight 1780
Physical Form Granular Powder
Chemical Name or Material Melanin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.