Learn More
Bachem (Nle4,D-Phe7)-a-MSH Trifluoroacetate
SDP
Supplier:  Bachem 4003800.0001
Encompass_Preferred
(Nle4,D-Phe7)-a-MSH Trifluoroacetate, Ac-Ser-Tyr-Ser-Nle-Glu-His-D-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2, 75921-69-6, C78H111N21O19, Mr 1646.87, 1mg. NDP-MSH, Afamelanotide, Melanotan 1, CUV1647, [Nle4, DPhe7] a-MSH, amide. Afamelanotide, a synthetic analog of a-MSH which shows prolonged biological activity and is 26 times more active than a-MSH in the adenylate cyclase assay.
Catalog No. NC1037522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.