Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Melatonin, MT1 and MT2 Receptor Agonist, 99 Percent Purity, 50 mg
Description
- Purity: >99%
- Molecular Weight: 232.28 Da
- Form: Solid
- Activity: Agonist at melatonin MT1 and MT2 receptors
- Key Functions: Endogenous hormone important in the control of the circadian rhythm; possesses antioxidative and immunomodulatory effects
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | MT1 Receptor, MT2 Receptor |
| Product Type | Antioxidant |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% Melatonin |
| Quantity | 50 mg |
| For Use With (Application) | Circadian rhythm research, antioxidative effects, immunomodulatory effects |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?