Learn More
CPC Scientific H-Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-Trp-Ile-Lys-Arg-Lys-Arg-Gln-Gln-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP018B
Encompass
26-residue polypeptide isolated from bee venom that binds calmodulin in a Ca2+ dependent manner, activates phospholipase A2, and inhibits protein kinase C (IC50 = 5-7 m M) by binding to the catalytic domain in a Mg-ATP sensitive manner. IC50 of 1-4 µM.ONE-LETTER SEQUENCE: GIGAVLKVLTTGLPALISWIKRKRQQ-NH2MOLECULAR FORMULA: C131H229N39O31MOLECULAR WEIGHT:2846.46STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [20449-79-0], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Raynor et al., J. Biol. Chem., 266, 2753 (1991); Schweitz, Toxicon, 22, 308 (1984)
Catalog No. 50-211-0171
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.