Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melphalan, 95%, MP Biomedicals™

Catalog No. ICN15534583 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15534583 250 mg
ICN15534580 100 mg
ICN15534591 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15534583 Supplier MP Biomedicals, Inc Supplier No. 0215534583

Chemical Identifiers

CAS 148-82-3
Molecular Formula C13H18Cl2N2O2
Molecular Weight (g/mol) 305.199
InChI Key SGDBTWWWUNNDEQ-LBPRGKRZSA-N
Synonym melphalan, alkeran, melfalan, phenylalanine mustard, l-sarcolysine, l-pam, l-sarcolysin, l-phenylalanine mustard, levofalan, p-l-sarcolysin
PubChem CID 460612
ChEBI CHEBI:28876
IUPAC Name (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)N(CCCl)CCCl

Specifications

Color Yellow
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 305.2
Physical Form Powder
Chemical Name or Material Melphalan
Recommended Storage Store at room temperature.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.