Learn More
Electron Microscopy Sciences MEM-32 Grid Storage Box
SDP
Supplier:  Electron Microscopy Sciences 7116501
Encompass_Preferred
This multipurpose electron microscope specimen grid box eliminates the chances of tweezer insertion damaging the grids because the tweezers slot' and grid slot' are in a separate location. The tweezers are only able to grip the edge of the grid enabling it to be picked up. And the grids stored no longer jump out of the box while you remove the cover. Between the body of the box and the sliding lid, there is a separate plastic cover, which allows foronly four slots beingexposed at a time. The Grid Record Card is stored safely by insertion along on the reverse side of the box.
Catalog No. 50-930-877
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.