Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEM chloride, 94%

Catalog No. p-3736818
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC185980100 10 g Glass bottle
AC185980250 25 g Glass bottle
AC185981000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC185980100 Supplier Thermo Scientific Chemicals Supplier No. 185980100
Only null left
Add to Cart

CAS: 3970-21-6 | C4H9ClO2 | 124.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3970-21-6 , 542-88-1
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00000888
InChI Key BIAAQBNMRITRDV-UHFFFAOYSA-N
Synonym 2-methoxyethoxymethyl chloride, 1-chloromethoxy-2-methoxyethane, ethane, 1-chloromethoxy-2-methoxy, mem chloride, methoxyethoxymethyl chloride, mem-chloride, 1-chloro-2,5-dioxahexane, 2-methoxyethoxy methyl chloride, beta-methoxyethoxymethyl chloride, .beta.-methoxyethoxymethyl chloride
PubChem CID 77590
IUPAC Name 1-(chloromethoxy)-2-methoxyethane
SMILES COCCOCCl

Specifications

Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 50.0°C to 52.0°C (13.0 mmHg)
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.4260 to 1.4280
Linear Formula CH3OCH2CH2OCH2Cl
Quantity 10 g
Fieser 07,227; 09,299
Specific Gravity 1.09
Formula Weight 124.57
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material MEM chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause cancer.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 223-589-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.