Learn More
Apexbio Technology LLC Memantine hydrochloride, 1g Cas# 41100-52-1
SDP
Supplier:  Apexbio Technology LLC B33081000
Encompass_Preferred
NMDA receptor antagonist. Memantine hydrochloride. 1401033-86-0. MFCD28155269 Memantine hydrochloride is an antagonist of NMDA receptor with IC50 values of 0.93M, 0.82M and 0.47M for NRIa/NR2A, NRIa/NR2B and NRIa/NR2D, respectively [1]. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8700
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.