Learn More
Cytiva Versapor Acrylic Copolymer 1200 Membrane Disc Filter, 1.2 µm - 47 mm (100/pkg)
Catalog No. 502063316 Shop All Cytiva Products
Encompass
Ideal for prefiltration of difficult-to-filter solutions and serum.-For prefiltration-Excellent retention-Acrylic copolymerDesigned to be used in applications requiring excellent retention properties combined with tensile strengths in excess of 200 bar (3000 psi).Filter membrane is made of acrylic copolymer on a nonwoven support. Strong, flexible, and nonshedding, both dry and wet. Delivers high flow rates at low differential pressures. Can be used as a prefilter to replace glass fiber filters. UV or gamma sterilizable. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3316 Supplier Cytiva Supplier No. 66394
Ideal for prefiltration of difficult-to-filter solutions and serum.-For prefiltration-Excellent retention-Acrylic copolymerDesigned to be used in applications requiring excellent retention properties combined with tensile strengths in excess of 200 bar (3000 psi).Filter membrane is made of acrylic copolymer on a nonwoven support. Strong, flexible, and nonshedding, both dry and wet. Delivers high flow rates at low differential pressures. Can be used as a prefilter to replace glass fiber filters. UV or gamma sterilizable. This product is not approved for use in medical, clinical, surgical or other patient applications.