Learn More
Invitrogen™ Membrane Blocking Solution
Catalog No. 000105
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
00-010-5 1 L
1 options
Catalog No. 00-010-5 Supplier Invitrogen™ Supplier No. 000105
  • $259.00
    $194.65 / Each of 1
    Save $64.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Membrane Blocking Solution
Membrane Blocking Solution can be used to block nitrocellulose and PVDF membranes, as well as to dilute antibodies or enzyme conjugates. This solution is a Tris buffer containing bovine serum albumin, goat IgG, Tween 20, and a mixture of other components that can reduce non-specific binding. The Proclin used in this reagent does not inhibit peroxidase or alkaline phosphatase activity. Note that this blocking solution contains normal goat IgG and should not be used in assays where anti-goat immunoglobulins are employed.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Membrane Blocking Solution
Quantity 1 L
How can I reduce background bands in my Western blot?
Optimize the concentration of primary and secondary antibodies. In some cases, increasing the concentration of blocking agent (BSA or non-fat dry milk) or usiing an alternative blocking solution such as Starting Block or SuperBlock may reduce background signal. After incubation with the primary antibody, wash at least 2 times with TBST (include 0.5 M NaCl in one or more of the wash steps). Avoid Nonidet P40 or Triton X-100 in buffers because protein detection is decreased when these detergents are used.
For Research Use Only. Not for use in diagnostic procedures.