Learn More
Fisherbrand™ Membrane Filter, 0.45 μm
Catalog No. FB14955206 Shop All Fisherbrand Products
Change view
Click to view available options
Membrane Filter, 0.45 μm
Catalog No.
FB14955206 Membrane Filter, 0.45 μm
1 options
Catalog No. FB14955206
Add to Cart
Add to Cart
  • For use with the Fisherbrand Pipet Controller
  • 0.45 μm Hydrophobic PTFE
  • Pack of 5 replacement Membrane Filters
Specifications
Description Replacement parts only
Material PTFE
Volume (Metric) 0.45 μm
For Use With (Equipment) Fisherbrand Pipet Controller (FB14955202)
Quantity 5 Pieces