Learn More
Memmert Memmert Constant Climate Chamber HPP750Eco. 750L - 115V, Stainless Steel Construction with TwinDISPLAY digital controller
SDP
Supplier:  Memmert HPP750ECO 115V
Encompass_Preferred
Memmert Constant Climate Chamber (HPPeco) - this programmable model is the 115 volt, Peltier-technology seamlessly heats and cools, with TwinDISPLAY digital controller. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Atmocontrol Manual, Operation manual, Calibration Certificate and Power Cord - energy efficient, precise. Setting temperature range +5C to +70C. Setting humidity range 10%RH - 90%RH. Internal dimensions 1040/1200/568-W/H/D mm.
Catalog No. 50-225-4407
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.