Learn More
Memmert Memmert Incubator IF750. 750L - 230V, Forced-Air Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Supplier:  Memmert IF750 230V
Encompass_Preferred
Memmert Incubators - this model is the 230 volt, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 1040/1200/561-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient up to +80C.
Catalog No. 50-225-4370
Only null left
Add to Cart
Request a Quote