Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UN260. 260L-115 Volt, Natural Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
Supplier: Memmert UN260
Memmert Universal heating and drying Oven - this model is the 115 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 640/800/500-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient to +300C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.