Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UN30Plus. 30L - Programmable Model - 115 Volt, Natural Convection, Stainless Steel Construction with TwinDISPLAY Digital Controller.
SDP
Supplier:  Memmert UN30 PLUS
Encompass_Preferred
Memmert Universal heating and drying Oven - this programmable model is the 115 volt, natural convection, with TwinDISPLAY digital controller. Interior dimensions 400/320/250-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Atmocontrol Software, Manual, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient to +300C.
Catalog No. 50-225-4247
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.