Learn More
Memmert Memmert Incubator IN55. 55L - 115V, Natural Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Supplier:  Memmert IN55
Encompass_Preferred
Memmert Incubators - this model is the 115 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 400/400/330-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient up to +80C.
Catalog No. 50-225-4313
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.