Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cut Pro™ 92723PU HyperMax™ Gloves, Black Polyurethane Coating

Catalog No. 19812610 Shop All MCR Safety Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-610 X-Small
19-812-611 Small
19-812-612 Medium
19-812-614 Large
19-812-615 X-Large
19-812-616 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-812-610 Supplier MCR Safety Supplier No. 92723PUXS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Salt and pepper colored HyperMax™ shell features black polyurethane coating on palm and fingertips for cut, tear and abrasion resistance.

  • Salt and pepper colored 13-gauge HyperMax™ shell
  • Black polyurethane coated palm and fingertips
  • Seamless shell is comfortable, cool and easy to wear

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI Cut 2
Cuff Material HPPE
Coating Material Polyurethane
Color Black, Multicolored
Cut Resistance Rating ANSI A2
Material HPPE
Product Type Cut-resistant Gloves
Size X-Small
For Use With (Application) Assembly Line Production, Contruction, Material Handling
Glove Cuff Color Multi-colored
Glove Cuff Style Knit
Hem Color Multi-colored
Model 92723PUXS
Series Cut Pro™ 92723PU
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.