Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cut Pro™ 92733PU Black HyperMax™ Gloves, Polyurethane Coating

Catalog No. 19812620 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-620 Large
19-812-617 Small
19-812-619 Medium
19-812-621 X-Large
19-812-622 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-620 Supplier MCR Safety Supplier No. 92733PUL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Black HyperMax™ shell features polyurethane coating on the palm and fingertips for cut, tear and abrasion resistance. Black color hides dirt and grease.

  • 13-gauge HPPE/Synthetic shell
  • Black polyurethane (PU) coated palm and fingertips
  • Abrasion, cut and puncture resistant
  • Overall black color hides soiling
  • Seamless shell is comfortable, cool and easy to wear

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A3
Material HPPE
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Assembly Line Production, Material Handling, Warehousing
Glove Cuff Style Knit
Hem Color Multi-colored
Model 92733PUL
Series Cut Pro™ 92733PU
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.