Learn More
Bulwark™ Men's Charcoal Plaid Flame Resistant Flex Knit Button Down Shirt
Shop All Red Kap Industries ProductsDescription
- The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
- Spandex-blend fabric allows it to stretch freely.
- Button-down collar keeps you looking crisp and sharp all day.
- Easy-to-spot CAT 2/2112 label on the front pocket and left sleeve leave no question that you're protected on the job site.
- Red buttonhole stitching and right sleeve placket leave no doubt you're armed with the very best.
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 1
- Primary Closure: Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
- Blend: Body: 93% Cotton / 7% Spandex Elastane
- Features: Button down collar, Single chest pocket with button closure, Lightweight, stretch fabric
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
- Gender: Male
- Brand: Bulwark™
Specifications
Specifications
| Size | 2X-Large |
| Gender | Men's |
| Material | Cotton, Spandex |
| Color | Charcoal |
| Neck Style | Collar |
| Sleeve Style | Long Sleeve |
| Construction | Knit |
| Closure Type | Button |
| Disposable | Reusable |
| No. of Pockets | 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.