Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Men's Stretch Denim Flame Resistant Dungaree

Catalog No. 17204449 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
36 x 30 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-204-449 36 x 30 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-204-449 Supplier Bulwark™ Supplier No. PSJ6SD 36 30
Only null left
Add to Cart
Add to Cart

Men's Stretch Denim Flame Resistant Dungaree fits like your weekend jeans and moves with you while you work. Relaxed through waist, hip, and thigh to allow for easy movement.

  • Increased durability with triple needle felled out seams and seat seam
  • Sits below natural waist with relaxed fit through waist, hip, and thigh
  • Bootcut leg opening fits easily over boots
  • Five traditional jean style pockets, with an additional ruler pocket on right leg and utility pocket on left leg for extra storage
  • One-piece waistband with metal button closure (metal is not exposed to skin)
  • Utility hammer loop on left hip
  • Reinforced pockets help prevent holes
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 7
  • Primary Closure: Concealed Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 12 oz. (405 g/m2) Flame Resistant Denim
  • Blend: Body: 94% Cotton/5% Polyester/1% Spandex Elastane
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size 36 x 30 in.
Gender Men's
Material Cotton, Polyester, Spandex
Color Denim
Seam Type Stitched
No. of Pockets 6
Closure Type Concealed Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 15
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.