Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terra™ Men's Terra Patton 6 in. Aluminum Toe Safety Work Boot

Take on tough jobs in absolute comfort - designed to keep the weight down without compromising on safety. The aluminum safety toe gives you the protection you need, while molded TPU heel and toe bumpers fend off scuffs.

Supplier:  Terra™ 4NS6BN 085 W

 View more versions of this product

Catalog No. 17-100-360


Only null left
Add to Cart

Description

Description

  • Premium, full grain, lightly pebbled, waterproof leather upper
  • Traditional padded top collar
  • Moisture-wicking lining with CLEANFEET™ odor-control treatment
  • Molded TPU toe bumper for increased abrasion resistance
  • Cemented, shock absorbing, high rebound PU-Dual-density high rebound EVA JETBOUND™ FOOTBED™ insole midsole
Specifications

Specifications

Brown
8.5W
No
Leather
Men's
Boot
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.