Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menthol, Crystal, FCC, Spectrum™ Chemical
SDP

Catalog No. 18604299 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-299 125 g Amber Glass Bottle
18-604-298 25 g Amber Glass Bottle
18-604-300 500 g Amber Glass Bottle
18-604-301 12 kg Amber Glass Bottle
18-604-302 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-299 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1360125GM
Add to Cart
Add to Cart

M1360, 2216-51-5, C10H20O

Spectrum™ Chemical Menthol, Crystal, FCC is used as a flavorant in the food industry. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
IUPAC Name (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

CAS Max % 100%
Melting Range 41-44°C
Packaging Amber Glass Bottle
Quantity 125 g
Specific Rotation -45° to -51°
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Menthol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.