Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Menthyl Chloroformate 97.0+%, TCI America™
SDP

Catalog No. M099025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M099025ML 25 mL
M09905ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M099025ML Supplier TCI America Supplier No. M099025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14602-86-9
Molecular Formula C11H19ClO2
Molecular Weight (g/mol) 218.72
MDL Number MFCD00009694,MFCD00190089
InChI Key KIUPCUCGVCGPPA-UHFFFAOYNA-N
Synonym 1r---menthyl chloroformate, --menthyl chloroformate, 1r,2s,5r-2-isopropyl-5-methylcyclohexyl carbonochloridate, --menthylchloroformate, pubchem8110, ?-menthyl chloroformate, --chloroformyloxy-p-menthane, 1r,2s,5r-2-isopropyl-5-methylcyclohexylcarbonochloridate, 1r,2s,5r-5-methyl-2-propan-2-yl cyclohexyl chloroformate, 1r---menthyl chloroformate, optical purity ee glc
PubChem CID 7014897
IUPAC Name 5-methyl-2-(propan-2-yl)cyclohexyl carbonochloridate
SMILES CC(C)C1CCC(C)CC1OC(Cl)=O

Specifications

Color Yellow
Boiling Point 109°C
Quantity 25 mL
UN Number 3277
Formula Weight 218.72
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material (-)-Menthyl Chloroformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.