Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEPCE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562438 Supplier Invitrogen™ Supplier No. PA562438
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQTPEYDVVL CLSLTKWVHL NWGDEGLKRM FRRIYRHLRP GGILVLEPQP WSSYGKRKTL TETIYKNYYR IQLKPEQFSS YLTSPDVGFS SYELVATPHN TSKGFQRPVY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

S-adenosyl-L-methionine-dependent methyltransferase that adds a methylphosphate cap at the 5'-end of 7SK snRNA (7SK RNA), leading to stabilize it (PubMed:17643375, PubMed:19906723, PubMed:30559425). Also has a non-enzymatic function as part of the 7SK RNP complex: the 7SK RNP complex sequesters the positive transcription elongation factor b (P-TEFb) in a large inactive 7SK RNP complex preventing RNA polymerase II phosphorylation and subsequent transcriptional elongation (PubMed:17643375). The 7SK RNP complex also promotes snRNA gene transcription by RNA polymerase II via interaction with the little elongation complex (LEC) (PubMed:28254838). In the 7SK RNP complex, MEPCE is required to stabilize 7SK RNA and facilitate the assembly of 7SK RNP complex (PubMed:19906723). MEPCE has a non-enzymatic function in the 7SK RNP complex; interaction with LARP7 within the 7SK RNP complex occluding its catalytic center (PubMed:19906723). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEPCE
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene mepce
Gene Accession No. Q7L2J0
Gene Alias 7SK snRNA methylphosphate capping enzyme; BCDIN3; Bicoid-interacting protein 3 homolog; bin3 homolog; bin3, bicoid-interacting 3; bin3, bicoid-interacting 3, homolog; Bipl1; D5Wsu46e; fc59f06; FLJ20257; Mepce; methylphosphate capping enzyme; si:ch211-283h6.1; wu:fc59f06; zBCDIN3
Gene Symbols mepce
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEPCE. Recombinant protein control fragment (Product #RP-92757).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.