Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEPCE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7206237 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9833 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9833 Supplier Proteintech Group Inc Supplier No. 149171AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S-adenosyl-L-methionine-dependent methyltransferase that adds a methylphosphate cap at the 5′-end of 7SK snRNA, leading to stabilize it.

Specifications

Antigen MEPCE
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MEPCE
Gene Accession No. Q7L2J0, Q8K3A9
Gene Alias Bin3 homolog, MEPCE, methylphosphate capping enzyme
Gene Symbols MEPCE
Host Species Rabbit
Immunogen MEPCE Fusion Protein Ag6733
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231803, 304361, 56257
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.