Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mepivacaine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M25601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25601G 1 g
M25605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25601G Supplier TCI America Supplier No. M25601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1722-62-9
Molecular Formula C15H23ClN2O
Molecular Weight (g/mol) 282.812
MDL Number MFCD00144738
InChI Key RETIMRUQNCDCQB-UHFFFAOYSA-N
Synonym N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride
PubChem CID 66070
ChEBI CHEBI:6760
IUPAC Name N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide;hydrochloride
SMILES CC1=C(C(=CC=C1)C)NC(=O)C2CCCCN2C.Cl

Specifications

Melting Point 263°C
Color White
Quantity 1 g
Formula Weight 282.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mepivacaine Hydrochloride
RTECSNumber TK6475000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.