Learn More
Thermo Scientific™ MEPS™ for GC: CTC Analytics Using 250¦L Syringes
Catalog No. 60308305
Change view
Click to view available options
Description:
C18 MEPS for GC; For CTC
Hypercarb MEPS for GC; For CTC
MEPS Development Kit for GC
Retain PEP MEPS for GC; For CTC
Retain-AX MEPS for GC; For CTC
Retain-CX MEPS for GC; For CTC
Silica MEPS for GC; For CTC
Verify-AX MEPS for GC; For CTC
Verify-CX MEPS for GC; For CTC
Sorbent:
HyperSep C18 MEPS
HyperSep Hypercarb MEPS
HyperSep Retain AX MEPS
HyperSep Retain CX MEPS
HyperSep Retain PEP MEPS
HyperSep Retain PEP, Retain CX, Retain AX, Hypercarb, and C18
HyperSep Silica MEPS
HyperSep Verify AX MEPS
HyperSep Verify CX MEPS
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Sorbent
60-308-305 Verify-CX MEPS for GC; For CTC HyperSep Verify CX MEPS
60-308-301 Retain PEP MEPS for GC; For CTC HyperSep Retain PEP MEPS
60-308-302 Retain-CX MEPS for GC; For CTC HyperSep Retain CX MEPS
60-308-303 Retain-AX MEPS for GC; For CTC HyperSep Retain AX MEPS
60-308-304 Hypercarb MEPS for GC; For CTC HyperSep Hypercarb MEPS
60-308-306 Verify-AX MEPS for GC; For CTC HyperSep Verify AX MEPS
60-308-307 C18 MEPS for GC; For CTC HyperSep C18 MEPS
60-308-308 Silica MEPS for GC; For CTC HyperSep Silica MEPS
60-308-309 MEPS Development Kit for GC HyperSep Retain PEP, Retain CX, Retain AX, Hypercarb, and C18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60-308-305 Supplier Thermo Scientific™ Supplier No. 60308305
Save hours in sample preparation, and go from extraction to injection in a single process with these 250μL syringes for GC applications using CTC Analytics.
Save hours in sample preparation, and go from extraction to injection in a single process with Thermo Scientific™ HyperSep™ MEPS™ (Micro Extraction by Packed Sorbent) technology.
Suitable for reversed phases, normal phases, mixed mode or ion exchange chemistries, Thermo Scientific HyperSep MEPS Syringes and Components for Analytics are available in a variety of SPE phase chemistries. The HyperSep MEPS Barrel Insert and Needle Assembly (BIN) contains the stationary phase, and is built into the syringe needle.
These HyperSep MEPS BINs are designed for use with Thermo Scientific, CTC Analytics, HTA and Varian 8400 GC Systems.
MEPS performs the same functions as SPE—the removal of interfering matrix components and the selective isolation and concentration of analytes—except that it miniaturizes conventional SPE packed bed devices from milliliter bed volumes to microliter volumes, and incorporates them right into the syringe.
Mechanically robust, the technique offers significant advantages over conventional SPE extraction for method development and other GC applications:
Significantly reduces the time to prepare and inject samples from hours to minutes—typically less than two minutes per sample
Eliminates all extra steps between sample preparation and sample injection; the technique is fully compatible with direct injection onto GC
Reduces buffer and solvent volume from milliliters to microliters
Reduces the required sample volume to as little as 3.6µL, making the techniques especially well-suited for on-line use with volume-limited samples
Can easily be automated compatible and is compatible with CTC and eVol platforms
TRUSTED_SUSTAINABILITY
Specifications
Description Verify-CX MEPS for GC; For CTC
For Use With (Application) Gas Chromatography
For Use With (Equipment) CTC Analytics Systems Using 250 μL Syringes
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep Verify CX MEPS
Type MEPS Barrel Insert and Needle (BIN)