Learn More
AdipoGen Merafloxacin
SDP
Supplier:  AdipoGen AGCR13756M025
Encompass_Preferred
Chemical. CAS 91188-00-0. Formula C19H23F2N3O3. MW 379.4. Merafloxacin is a fluoroquinolone broad-spectrum antibacterial compound. It is an inhibitor of bacterial DNA gyrase and Type II DNA topoisomerase. It demonstrated excellent activity against Gram-positive organisms and less potent activity against Gram-negative bacteria. Merafloxacin is a programmed -1 ribosomal frameshifting -1 PRF inhibitor of SARS-CoV-2. Frameshift inhibition by merafloxacin is robust to mutations within the pseudoknot region and is similarly effective on -1 PRF of other betacoronaviruses and blocks SARS-CoV-2 replication in Vero E6 cells. The compound reduced viral levels in infected African green monkey VeroE6 cells in a concentration-dependent manner. Merafloxacin showed no cellular toxicity and resulted in a 3 to 4 orders of magnitude reduction of SARS-CoV-2 titer, with IC50 of 4.3 µMu.
Catalog No. 50-256-2169
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.