Learn More
Thermco Products Inc Precision lab Hg thermometer, 0 deg./230 deg.F, 2 deg.F div, 76mm imm, 305mm
Supplier: Thermco Products Inc ACC6103
High precision mercury SAMA thermometer that meet or exceed the specifications of the American National Standards Institute. Specifications approve by N.I.S.T. and ASTM. Individually serialized and permanetly marked. Made with precision yellow backed tubing. SAMA No. FP15
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.