Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Mercaptoethylamine hydrochloride, ∽98%, MP Biomedicals™

Catalog No. ICN10099525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10099525 25 g
ICN10099505 5 g
ICN10099580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10099525 Supplier MP Biomedicals, Inc Supplier No. 0210099525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
SMILES [H+].[Cl-].NCCS

Specifications

Color White
Assay Percent Range ∽ 98%
Quantity 25 g
IUPAC Name hydrogen 2-aminoethane-1-thiol chloride
Formula Weight 113.6
Physical Form Powder
Chemical Name or Material β-Mercaptoethylamine hydrochloride
Recommended Storage Store at 4°C, dessicated.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.