Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Acetate, Powder, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18611531 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-531 500 g Amber Glass Bottle
18-611-525 25 g Amber Glass Bottle
18-611-527 125 g Amber Glass Bottle
18-611-529 2.5 kg Amber Glass Bottle
18-611-528 12 kg Poly Pail
18-611-530 25 kg Fiber Drum
18-611-532 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-611-531 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1140500GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
IUPAC Name mercury(2+) diacetate
SMILES [Hg++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 98%
Grade ACS
Packaging Amber Glass Bottle

Caution: for manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.