Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Nitrate, (1mL = 5mg Cl-), for Chloride, Certified, 0.141N ±0.002N (0.071M), LabChem™

Catalog No. LC166601
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC166601 500 mL
LC166602 1 L
LC166604 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC166601 Supplier LabChem, Inc. Supplier No. LC166601
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7783-34-8 , 7697-37-2 , 7732-18-5
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
IUPAC Name mercury(2+) hydrate dinitrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Colorless
CAS Max % 2.54, 96.46, 1
Linear Formula Hg(NO3)2·H2O
Concentration 0.141N ±0.002N (0.071M)
Density 1g/mL
Solubility Information Soluble in water
PubChem CID 91886652  
Formula Weight 342.62
Grade Certified
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Nitrogen oxides; Mercury
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Mercuric Nitrate, (1mL = 5mg Cl-), for Chloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed or if inhaled.
Causes skin irritation and serious eye irritation.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.