accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOONOOHgH2O

CAS RN 7783-34-8

IUPAC Name: mercury(2+) hydrate dinitrate
Synonyms: mercuric nitrate monohydrate
Molecular Weight (g/mol): 342.62
Molecular Formula: H2HgN2O7
InChi Key: KVICROHOONHSRH-UHFFFAOYSA-N
SMILES: O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (10)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (6)
Grade 
  • (2)
  • (3)
  • (2)
  • (6)
  • (5)
  • (17)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
Physical Form 
  • (2)
  • (5)
  • (2)
  • (6)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (2)
  • (6)
  • (4)
  • (1)
  • (18)
  • (6)
  • (8)
  • (1)
  • (10)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (6)
  • (2)
  • (3)
  • (2)
  • (6)
  • (5)
  • (17)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (2)
  • (5)
  • (2)
  • (6)
  • (1)
  • (2)
  • (2)