Learn More
Mercuric Nitrate, (1mL = 5mg Cl-), for Chloride, Certified, 0.141N ±0.002N (0.071M), LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| Color | Colorless |
| CAS Max % | 1, 96.46, 2.54 |
| Linear Formula | Hg(NO3)2·H2O |
| Concentration | 0.141N ±0.002N (0.071M) |
| Density | 1g/mL |
| Solubility Information | Soluble in water |
| PubChem CID | 944 |
| Formula Weight | 342.62 |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed or if inhaled. Causes skin irritation and serious eye irritation. May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Do not eat, drink or smoke when using this product. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin after handling. Use only outdoors or in a well-ventilated area. Avoid release to the environment. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. If on skin: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. Take off contaminated clothing and wash before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Collect spillage. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.