Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Nitrate, Monohydrate, ACS Grade, ≥ 98.0 , LabChem™

Catalog No. LC166459 Shop All LabChem Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC166459 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC166459 Supplier LabChem, Inc. Supplier No. LC166459
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7783-34-8
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+) hydrate dinitrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 70°C
Color White
Physical Form Crystals
CAS Max % 100
Quantity 100 g
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 4.3g/cm3
Decomposition Information Mercury; Nitrogen oxides
Density 4.3g/cm3
Linear Formula Hg(NO3)2·H2O
UN Number UN1625
Solubility Information Soluble in water
Formula Weight 342.62
Percent Purity ≥98.0%
Grade ACS
Assay as Hg(NO3)2·H2O
Chemical Name or Material Mercuric Nitrate, Monohydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Fatal if swallowed, in contact with skin or if inhaled.
May cause damage to organs (kidney, central nervous system) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe dust.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin: Wash with plenty of soap and water.
Immediately call a poison center/doctor.
Remove immediately all contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.