Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Oxide, Red, Powder, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604316 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-316 2.5 kg Amber Glass Bottle
18-604-313 25 g Amber Glass Bottle
18-604-314 125 g Amber Glass Bottle
18-604-315 500 g Amber Glass Bottle
18-604-317 12 kg Poly Pail
18-604-318 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-316 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M11702.5KG
Add to Cart
Add to Cart

M1170, 21908-53-2, HgO

Spectrum™ Chemical Mercuric Oxide, Red, Powder, Reagent, ACS , also known as mercury(II) oxide, is used to produce mercury as it decomposes quite easily and as a material for cathodes for mercury batteries. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

CAS Max % 100%
Quantity 2.5 kg
UN Number UN1641
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.