Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercurous Nitrate TS, Ricca Chemical
SDP

Catalog No. R4788000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4788000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. R4788000500 Supplier Ricca Chemical Company Supplier No. R4788000500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 7732-18-5 , 14836-60-3 , 7697-37-2 , 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Colorless
CAS Min % 84.0, 12.62, 1.33
CAS Max % 87.0, 13.13, 1.38
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Mercurous Nitrate, Test Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.