Learn More
New Pig Mercury Absorbent Powder
SDP
Catalog No. 18389142 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Mercury Absorbent Powder
Catalog No.
18-389-142 Type: Mercury Absorbent Powder
1 options
This item is not returnable. View return policy
Catalog No. 18-389-142 Supplier New Pig Supplier No. PLP602
Add to Cart
Add to Cart
This item is not returnable. View return policy
Binds to mercury to make the droplets easier to gather while it suppresses vapors for safer cleanup.
  • Sprinkle onto mercury to absorb up to 1,000 grams (35.27 ounces) and transform it into a zinc amalgam
  • Suppresses hazardous mercury vapors to limit worker exposure
  • Easy-to-handle shaker bottle lets you apply powder right where you need it
  • Ideal for removing mercury from work surfaces, benchtops, floors, cracks and crevices