Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury, Electronic Grade, 99.9998% (metals basis)

Catalog No. aa10242p2
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
10 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10242P2 1 lb.
AA10242P5 5 lb.
AA10242P9 10 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AA10242P2 Supplier Thermo Scientific Chemicals Supplier No. 010242P2
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 7439-97-6 | Hg | 200.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-97-6
Molecular Formula Hg
Molecular Weight (g/mol) 200.59
MDL Number MFCD00011035
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
PubChem CID 23931
ChEBI CHEBI:16170
IUPAC Name mercury
SMILES [Hg]

Specifications

Color Silver
Appearance Silver-colored liquid
Quantity 1 lb.
Odor Odorless
Form Liquid
Assay Percent Range 99.9998% (Metals basis)
UN Number UN2809
Synonym hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
Percent Purity 99.9998%
Grade Electronic
Assay (metals basis)
Chemical Name or Material Mercury
Show More Show Less
Hazard Category H330-H360D-H372
Hazard Statement GHS H Statement
H330-H360-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: MERCURY
EINECSNumber 231-106-7
RTECSNumber OV4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.