Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(I) chloride, 99+%, extra pure

Catalog No. p-3756669
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213120050 5 g Plastic bottle
AC213121000 100 g Plastic bottle
AC213125000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC213120050 Supplier Thermo Scientific Chemicals Supplier No. 213120050
Only null left
Add to Cart

CAS: 10112-91-1 | Cl2Hg2 | 472.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10112-91-1
Molecular Formula Cl2Hg2
Molecular Weight (g/mol) 472.08
MDL Number MFCD00011043
InChI Key ZOMNIUBKTOKEHS-UHFFFAOYSA-L
Synonym mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
PubChem CID 24956
ChEBI CHEBI:33050
IUPAC Name chloromercury
SMILES Cl[Hg].Cl[Hg]

Specifications

Melting Point 400.0°C
Color White
Physical Form Amorphous Powder
Quantity 5 g
Assay Percent Range 99% min. (Iodimetry)
Linear Formula Hg2Cl2
Merck Index 15, 5962
Solubility Information Solubility in water: practically insoluble. Other solubilities: insoluble in ether and alcohol
Formula Weight 472.08
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Density 7.15g/mL
Chemical Name or Material Mercury(I) chloride, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor
EINECSNumber 233-307-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.