CAS RN 10112-91-1
Mercury(I) chloride, 99.5%
CAS: 10112-91-1 Molecular Formula: Cl2Hg2 Molecular Weight (g/mol): 472.084 MDL Number: MFCD00011043 InChI Key: ZOMNIUBKTOKEHS-UHFFFAOYSA-L Synonym: mercurous chloride,calomel,mercury i chloride,mercury chloride hg2cl2,mercury subchloride,dimercury dichloride,calogreen,calotab,chlorure mercureux,ccris 3803 PubChem CID: 24956 ChEBI: CHEBI:33050 IUPAC Name: chloromercury SMILES: Cl[Hg].Cl[Hg]
Mercurous Chloride, Powder, ACS, 99.5%, Spectrum™ Chemical
CAS: 10112-91-1 Molecular Formula: Cl2Hg2 Molecular Weight (g/mol): 472.08 InChI Key: ZOMNIUBKTOKEHS-UHFFFAOYSA-L IUPAC Name: dimercuranebis(ylium) dichloride SMILES: [Cl-].[Cl-].[Hg+][Hg+]
Mercury(I) chloride, ACS reagent
CAS: 10112-91-1 Molecular Formula: Cl2Hg2 Molecular Weight (g/mol): 472.08 InChI Key: ZOMNIUBKTOKEHS-UHFFFAOYSA-L Synonym: mercurous chloride,calomel,mercury i chloride,mercury chloride hg2cl2,mercury subchloride,dimercury dichloride,calogreen,calotab,chlorure mercureux,ccris 3803 PubChem CID: 24956 ChEBI: CHEBI:33050 IUPAC Name: chloromercury SMILES: Cl[Hg].Cl[Hg]
Mercury(I) chloride, ACS, 99.5% min, Thermo Scientific Chemicals
CAS: 10112-91-1 Molecular Formula: Cl2Hg2 Molecular Weight (g/mol): 472.084 MDL Number: MFCD00011043 InChI Key: ZOMNIUBKTOKEHS-UHFFFAOYSA-L Synonym: mercurous chloride,calomel,mercury i chloride,mercury chloride hg2cl2,mercury subchloride,dimercury dichloride,calogreen,calotab,chlorure mercureux,ccris 3803 PubChem CID: 24956 ChEBI: CHEBI:33050 IUPAC Name: chloromercury SMILES: Cl[Hg].Cl[Hg]
Mercury(I) chloride, 99+%, extra pure
CAS: 10112-91-1 Molecular Formula: Cl2Hg2 Molecular Weight (g/mol): 472.08 MDL Number: MFCD00011043 InChI Key: ZOMNIUBKTOKEHS-UHFFFAOYSA-L Synonym: mercurous chloride,calomel,mercury i chloride,mercury chloride hg2cl2,mercury subchloride,dimercury dichloride,calogreen,calotab,chlorure mercureux,ccris 3803 PubChem CID: 24956 ChEBI: CHEBI:33050 IUPAC Name: chloromercury SMILES: Cl[Hg].Cl[Hg]
Mercury(I) Chloride, ≥99.5%, ACS Reagent, Solstice
CAS: 10112-91-1 Molecular Formula: Cl2Hg2 Molecular Weight (g/mol): 472.084 MDL Number: MFCD00011043 InChI Key: ZOMNIUBKTOKEHS-UHFFFAOYSA-L Synonym: mercurous chloride,calomel,mercury i chloride,mercury chloride hg2cl2,mercury subchloride,dimercury dichloride,calogreen,calotab,chlorure mercureux,ccris 3803 PubChem CID: 24956 ChEBI: CHEBI:33050 IUPAC Name: chloromercury SMILES: Cl[Hg].Cl[Hg]
Mercury(I) chloride, 99%, Thermo Scientific™
CAS: 10112-91-1 Molecular Formula: Cl2Hg2 Molecular Weight (g/mol): 472.084 MDL Number: MFCD00011043 InChI Key: ZOMNIUBKTOKEHS-UHFFFAOYSA-L Synonym: mercurous chloride,calomel,mercury i chloride,mercury chloride hg2cl2,mercury subchloride,dimercury dichloride,calogreen,calotab,chlorure mercureux,ccris 3803 PubChem CID: 24956 ChEBI: CHEBI:33050 IUPAC Name: chloromercury SMILES: Cl[Hg].Cl[Hg]