Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(I) chloride, 99%, Thermo Scientific™

Catalog No. AAA1249922
Change view
Click to view available options
Quantity:
100 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1249922 100 g
AAA124990E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1249922 Supplier Thermo Scientific Chemicals Supplier No. A1249922

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10112-91-1
Molecular Formula Cl2Hg2
Molecular Weight (g/mol) 472.084
MDL Number MFCD00011043
InChI Key ZOMNIUBKTOKEHS-UHFFFAOYSA-L
Synonym mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
PubChem CID 24956
ChEBI CHEBI:33050
IUPAC Name chloromercury
SMILES Cl[Hg].Cl[Hg]

Specifications

Quantity 100 g
UN Number UN2025
Formula Weight 472.09
Percent Purity 99%
Odor Odorless
Chemical Name or Material Mercury(I) chloride
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.