Learn More
Mercury(I) nitrate dihydrate, 98%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 70.0°C |
| Color | White to Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Assay Percent Range | 97.5% min |
| Linear Formula | Hg2(NO3)2·2H2O |
| Fieser | 06,360 |
| Merck Index | 13, 5923 |
| Solubility Information | Solubility in water: 20g/L (20°C). |
| Formula Weight | 561.22 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life with long lasting effects. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash w |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.