Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(I) nitrate dihydrate, 97%

Catalog No. AAA162680B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA162680B 1000 g
AAA1626818 50 g
AAA1626830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA162680B Supplier Thermo Scientific Chemicals Supplier No. A162680B
Add to Cart

CAS: 7782-86-7 | Hg2(NO3)2·2H2O

Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with alcohols, sulfur and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-86-7
Molecular Formula Hg2(NO3)2·2H2O
MDL Number MFCD00150812
Synonym Mercurous nitrate

Specifications

Melting Point 70°C
Quantity 1000 g
Linear Formula Hg2(NO3)2·2H2O
UN Number UN1627
Merck Index 14,5896
Solubility Information Slightly soluble in water.
Formula Weight 561.22 (525.19 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Light sensitive
Density 4.79
Chemical Name or Material Mercury(I) nitrate dihydrate
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCUROUS NITRATE
EINECSNumber 233-886-4
RTECSNumber OW8000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.