Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(Ii) Acetate, ≥99.0% (Precipitation Titration), Solstice

Catalog No. 60047652 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-652 250 g Poly bottle
60-047-653 50 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-652 Supplier Solstice Supplier No. 83352250G
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent,≥99.0% (precipitation titration)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
MDL Number MFCD00012165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Melting Point 179°C to 182°C
Color Beige
Physical Form Powder
Quantity 250 g
Linear Formula (CH3COO)2Hg
UN Number UN1629
Beilstein 3563831
Formula Weight 318.68g/mol
Percent Purity ≥99.0% (precipitation titration)
Odor Characteristic
Packaging Poly bottle
Chemical Name or Material Mercury(II) acetate
Show More Show Less
Hazard Statement P260-P273-P280-P301 + P310 + P330-P302 + P352 + P310-P304 + P340 + P310
DOTInformation 6.1 / PGII
RTECSNumber 216-491-1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.