Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) chloride, ACS, 99.5% min

Catalog No. AA1227436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1227436 500 g
AA1227414 25 g
AA1227422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1227436 Supplier Thermo Scientific Chemicals Supplier No. 01227436
Add to Cart

CAS: 7487-94-7 | Cl2Hg | 271.49 g/mol

Mercury(II) chloride is used as a catalyst in making plastics, depolarizer in batteries and as a reagent in organic synthesis and analytical chemistry. It is also used to prepare an amalgam with metals such as aluminum, which finds application as reducing agent in organic synthesis. It also finds application in photography as a photographic intensifier. Further, it serves as a preservative and used in the treatment of syphilis. It is also used to remove dithiane groups bonded to a carbonyl in an umpolung reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Boiling Point 302°C
Melting Point 280°C to 282°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5% min.
Solubility Information Soluble in boiling water (solubility in cold water is increased by HCl or alkali chlorides),alcohol,glycerol,methanol,acetone,and ethyl acetateSoluble in water,alcohol,acetone and ethyl acetate. Slightly soluble in benzene,carbon disulfide and pyridine.
Formula Weight 271.5
Grade ACS
Odor Odorless
Refractive Index 1.859
Appearance White powder
Vapor Pressure 0.0001 hPa
Density 5.4
Chemical Name or Material Mercury(II) chloride
Show More Show Less
Hazard Category H300-H310-H314-H335-H341-H361f-H372
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P201-P202-P260-P262-P264b-P270-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 231-299-8
RTECSNumber OV9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.