Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) nitrate monohydrate, ACS reagent

Catalog No. AC423945000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423945000 500 g Plastic bottle
AC423941000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423945000 Supplier Thermo Scientific Chemicals Supplier No. 423945000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 7783-34-8 | H2HgN2O7 | 342.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-34-8
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+) hydrate dinitrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 77.0°C to 79.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Linear Formula Hg(NO3)2·H2O
Fieser 03,197; 10,254; 11,317
Merck Index 14, 5880
Solubility Information Solubility in water: slightly soluble
Formula Weight 342.6
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 4.3
Density 4.3000g/mL
Chemical Name or Material Mercury(II) nitrate monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Re
Recommended Storage May become colored on exposure to light

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.