Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) Nitrate Solution, 0.05M, Solstice

Catalog No. 6003254 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-032-54 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-032-54 Supplier Solstice Supplier No. 342921L
Add to Cart
Add to Cart

Volumetric, 0.05 M Hg(NO3)2 (0.1N), for the determination of chloride

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-34-8
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+);dinitrate;hydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Hg+2]

Specifications

Quantity 1 L
Linear Formula Hg(NO3)2
UN Number UN3082
Formula Weight 324.6g/mol
Concentration or Composition (by Analyte or Components) 0.05M
Chemical Name or Material Mercury(II) nitrate solution
Hazard Statement P260-P280-P301 + P312 + P330-P302 + P352 + P312
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.