Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) sulfide, black, Puratronic™, 99.999% (metals basis)

Catalog No. AA1299206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1299206 5 g
AA1299214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1299206 Supplier Thermo Scientific Chemicals Supplier No. 01299206
Add to Cart

CAS: 1344-48-5 | HgS | 232.65 g/mol

Mercury(II) sulfide is used in paint, sealing wax and preservatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-48-5
Molecular Formula HgS
Molecular Weight (g/mol) 232.65
MDL Number MFCD00011046
InChI Key LGZXYFMMLRYXLK-UHFFFAOYSA-N
Synonym mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
PubChem CID 62402
IUPAC Name sulfanylidenemercury
SMILES [S--].[Hg++]

Specifications

Melting Point 583.5°C (sublimation)
Color Black
Physical Form Powder
Quantity 5 g
Merck Index 14,5888
Solubility Information Soluble in water (0.00001g/L).
Formula Weight 232.65
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Density 7.70 g/mL
Assay (metals basis)
Chemical Name or Material Mercury(II) sulfide
Show More Show Less
Hazard Category H300+H310+H330-H373
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 215-696-3
RTECSNumber OX0720000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.